COM448678397 Combi-Blocks, Inc.

3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid

  • MDL number MFCD04115349
  • Cas Number : 886362-90-9
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9859

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9859
Availability details for
Quantity
  • Availability
  • Add to Cart