COM448678401 Combi-Blocks, Inc.

3-[3-(2-Ethoxycarbonyl-ethoxy)-propoxy]-propionic acid

  • MDL number MFCD05863896
  • Cas Number : 886362-92-1
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9864
Availability details for
Quantity
  • Availability
  • Add to Cart