COM448678436 Combi-Blocks, Inc.

3-Acetylamino-3-(4-fluoro-phenyl)-propionic acid

  • MDL number MFCD01366670
  • Cas Number : 332052-58-1
  • Molecular Weight : 225.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9905
Availability details for
Quantity
  • Availability
  • Add to Cart