COM448678441 Combi-Blocks, Inc.

3-Amino-1h-pyrazolo[4,3-c]pyridin-4-ol

  • MDL number MFCD16877656
  • Cas Number : 900863-27-6
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9915
Availability details for
Quantity
  • Availability
  • Add to Cart