COM448678445 Combi-Blocks, Inc.

3-Amino-1-methyl-5-(methylthio)-1h-pyrazole-4-carbonitrile

  • MDL number MFCD02089753
  • Cas Number : 151291-05-3
  • Molecular Weight : 168.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9919
Availability details for
Quantity
  • Availability
  • Add to Cart