COM448678453 Combi-Blocks, Inc.

3-Amino-1-n-fmoc-piperidine, HCl

  • MDL number MFCD03001674
  • Cas Number : 811841-86-8
  • Molecular Weight : 358.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9927
Availability details for
Quantity
  • Availability
  • Add to Cart