COM448678478 Combi-Blocks, Inc.

3-Amino-2-cyclopentylpropionic acid

  • MDL number MFCD07372890
  • Cas Number : 910444-21-2
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9988
Availability details for
Quantity
  • Availability
  • Add to Cart