COM448678494 Combi-Blocks, Inc.

6-Bromo-2-chloronicotinonitrile

  • MDL number MFCD12401633
  • Cas Number : 1171919-79-1
  • Molecular Weight : 217.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0017
Availability details for
Quantity
  • Availability
  • Add to Cart