COM448678512 Combi-Blocks, Inc.

3-Amino-3-(2-amino-phenyl)-propan-1-ol

  • MDL number MFCD07784018
  • Cas Number : 886364-15-4
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0042

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0042
Availability details for
Quantity
  • Availability
  • Add to Cart