COM448678545 Combi-Blocks, Inc.

(3-Iodo-2,5,6-trimethoxypyridin-4-yl)boronic acid

  • MDL number MFCD22887387
  • Cas Number : 2096338-48-4
  • Molecular Weight : 338.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0096
Availability details for
Quantity
  • Availability
  • Add to Cart