COM448678569 Combi-Blocks, Inc.

3-Amino-3-(4'-cbz)piperidine-propionic acid ethyl ester

  • MDL number MFCD06796547
  • Cas Number : 886362-29-4
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0136
Availability details for
Quantity
  • Availability
  • Add to Cart