COM448678575 Combi-Blocks, Inc.

6-Chloro-3-iodoimidazo[1,2-a]pyridine-5-carbaldehyde

  • MDL number MFCD22887428
  • Cas Number : 1414864-07-5
  • Molecular Weight : 306.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0144
Availability details for
Quantity
  • Availability
  • Add to Cart