COM448678628 Combi-Blocks, Inc.

3-Amino-4,5-dimethyl-1h-pyrazole hoac

  • MDL number MFCD19382366
  • Cas Number : 116568-90-2
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0229
Availability details for
Quantity
  • Availability
  • Add to Cart