COM448678655 Combi-Blocks, Inc.

3-Amino-5-methyl-thiophene-2-carbonitrile

  • MDL number MFCD08689594
  • Cas Number : 83060-73-5
  • Molecular Weight : 138.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0279
Availability details for
Quantity
  • Availability
  • Add to Cart