COM448678674 Combi-Blocks, Inc.

3-Amino-7-methoxy-4h-chromen-4-one

  • MDL number MFCD17926186
  • Cas Number : 64915-37-3
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0309
Availability details for
Quantity
  • Availability
  • Add to Cart