COM448678706 Combi-Blocks, Inc.

3-Aminomethylpyridine-n-oxide

  • MDL number MFCD06212837
  • Cas Number : 10694-10-7
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0361
Availability details for
Quantity
  • Availability
  • Add to Cart