COM448678716 Combi-Blocks, Inc.

3-Amino-pentanoic acid, HCl

  • MDL number MFCD08450675
  • Cas Number : 80914-37-0
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0375
Availability details for
Quantity
  • Availability
  • Add to Cart