COM448678733 Combi-Blocks, Inc.

3-Amino-thiophene-2-carboxylic acid sodium salt

  • MDL number MFCD13194139
  • Cas Number : 78756-27-1
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0397
Availability details for
Quantity
  • Availability
  • Add to Cart