COM448678753 Combi-Blocks, Inc.

3-Benzoylindole

  • MDL number MFCD00022721
  • Cas Number : 15224-25-6
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0426
Availability details for
Quantity
  • Availability
  • Add to Cart