COM448678787 Combi-Blocks, Inc.

3-Biphenyl-3',4'-difluoro-acetic acid

  • MDL number MFCD03839980
  • Cas Number : 866108-76-1
  • Molecular Weight : 248.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0492
Availability details for
Quantity
  • Availability
  • Add to Cart