COM448678790 Combi-Blocks, Inc.

3-Boc-3-methylaminoazatidine

  • MDL number MFCD08061963
  • Cas Number : 577777-20-9
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0497
Availability details for
Quantity
  • Availability
  • Add to Cart