COM448678840 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-fluoroaniline

  • MDL number MFCD19382423
  • Cas Number : 1269232-95-2
  • Molecular Weight : 224.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0576
Availability details for
Quantity
  • Availability
  • Add to Cart