COM448678856 Combi-Blocks, Inc.

N-Benzyl-n-(2-methoxyethyl)amine

  • MDL number MFCD01445287
  • Cas Number : 51353-26-5
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0597
Availability details for
Quantity
  • Availability
  • Add to Cart