COM448678865 Combi-Blocks, Inc.

3-Bromomethyl-4-nitro-benzoic acid

  • MDL number MFCD08752706
  • Cas Number : 916791-27-0
  • Molecular Weight : 260.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0618
Availability details for
Quantity
  • Availability
  • Add to Cart