COM448678869 Combi-Blocks, Inc.

3-Bromo-N-toluidine HBr

  • MDL number MFCD09800390
  • Cas Number : 1159823-24-1
  • Molecular Weight : 266.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0626
Availability details for
Quantity
  • Availability
  • Add to Cart