COM448678894 Combi-Blocks, Inc.

3-Bromopyrazolo[1,5-a]pyridine-5-carboxylic acid

  • MDL number MFCD11518932
  • Cas Number : 876379-79-2
  • Molecular Weight : 241.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0659
Availability details for
Quantity
  • Availability
  • Add to Cart