COM448678930 Combi-Blocks, Inc.

(4-Methyl-4H-1,2,4-triazol-3-yl)methanamine

  • MDL number MFCD09863432
  • Cas Number : 145942-99-0
  • Molecular Weight : 112.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0719

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0719
Availability details for
Quantity
  • Availability
  • Add to Cart