COM448678938 Combi-Blocks, Inc.

3-Chloro-6-ethyl-pyridazine

  • MDL number MFCD08704287
  • Cas Number : 98198-61-9
  • Molecular Weight : 142.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0733
Availability details for
Quantity
  • Availability
  • Add to Cart