COM448678943 Combi-Blocks, Inc.

1-(5-Bromo-3-fluoropyridin-2-yl)piperazine

  • MDL number MFCD12198842
  • Cas Number : 1141669-85-3
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0743
Availability details for
Quantity
  • Availability
  • Add to Cart