COM448678950 Combi-Blocks, Inc.

3'-Chloromethylacetophenone

  • MDL number MFCD06658170
  • Cas Number : 41908-12-7
  • Molecular Weight : 168.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0752
Availability details for
Quantity
  • Availability
  • Add to Cart