COM448678951 Combi-Blocks, Inc.

N-(2-Chloroethyl)-n-methylbenzylamine, HCl

  • MDL number MFCD00034905
  • Cas Number : 23510-18-1
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0753
Availability details for
Quantity
  • Availability
  • Add to Cart