COM448678995 Combi-Blocks, Inc.

3-Cyclopentylpropionyl piperidine thioamide

  • MDL number MFCD02179405
  • Cas Number : 289677-08-3
  • Molecular Weight : 225.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0821
Availability details for
Quantity
  • Availability
  • Add to Cart