COM448679005 Combi-Blocks, Inc.

3-Ethoxycarbonylamino-5-boc-4,6-dihydro-pyrrolo[3,4-c]pyrazole

  • MDL number MFCD11053612
  • Cas Number : 1049677-66-8
  • Molecular Weight : 296.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0834
Availability details for
Quantity
  • Availability
  • Add to Cart