COM448679063 Combi-Blocks, Inc.

3H-Spiro[1-benzofuran-2,4'-piperidine]

  • MDL number MFCD09800537
  • Cas Number : 71916-73-9
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0925
Availability details for
Quantity
  • Availability
  • Add to Cart