COM448679087 Combi-Blocks, Inc.

3,4-Dihydro-2h-pyrano[3,2-b]pyridin-8-ol

  • MDL number MFCD22887431
  • Cas Number : 1414864-09-7
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0965
Availability details for
Quantity
  • Availability
  • Add to Cart