COM448679142 Combi-Blocks, Inc.

3-Methoxy-2,4,5-trifluorobenzonitrile

  • MDL number MFCD04115323
  • Cas Number : 112811-63-9
  • Molecular Weight : 187.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1051
Availability details for
Quantity
  • Availability
  • Add to Cart