COM448679145 Combi-Blocks, Inc.

3-Methoxy-2,4,5-trifluorobenzylamine

  • MDL number MFCD03839930
  • Cas Number : 886362-79-4
  • Molecular Weight : 191.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1056
Availability details for
Quantity
  • Availability
  • Add to Cart