COM448679246 Combi-Blocks, Inc.

Methoxymethyl 7-azaindole-5-carboxamide

  • MDL number MFCD18253848
  • Cas Number : 1246550-79-7
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1204
Availability details for
Quantity
  • Availability
  • Add to Cart