COM448679262 Combi-Blocks, Inc.

3-M-Tolyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD08685950
  • Cas Number : 886766-73-0
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1228
Availability details for
Quantity
  • Availability
  • Add to Cart