COM448679275 Combi-Blocks, Inc.

3-N-Cbz-aminomethylaniline hcl

  • MDL number MFCD09878799
  • Cas Number : 374554-26-4
  • Molecular Weight : 292.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1249
Availability details for
Quantity
  • Availability
  • Add to Cart