COM448679278 Combi-Blocks, Inc.

tert-Butyl ((6-chloroimidazo[1,2-a]pyridin-5-yl)methyl)carbamate

  • MDL number MFCD22887429
  • Cas Number : 1414864-04-2
  • Molecular Weight : 281.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1254
Availability details for
Quantity
  • Availability
  • Add to Cart