COM448679286 Combi-Blocks, Inc.

3-Nitro-2-methyl-benzylbromide

  • MDL number MFCD09745029
  • Cas Number : 39053-40-2
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1265
Availability details for
Quantity
  • Availability
  • Add to Cart