COM448679340 Combi-Blocks, Inc.

3-Phenylimidazo[1,2-a]pyrazine

  • MDL number MFCD18781863
  • Cas Number : 126052-34-4
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1343
Availability details for
Quantity
  • Availability
  • Add to Cart