COM448679342 Combi-Blocks, Inc.

3-Phenyl-pent-1-yn-3-ol

  • MDL number MFCD17926462
  • Cas Number : 6712-31-8
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1346
Availability details for
Quantity
  • Availability
  • Add to Cart