COM448679358 Combi-Blocks, Inc.

3-Pyrazol-1-yl-l-alanine 2hcl

  • MDL number MFCD09025315
  • Molecular Weight : 228.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1377
Availability details for
Quantity
  • Availability
  • Add to Cart