COM448679379 Combi-Blocks, Inc.

3-Pyrrolidin-1-ylacrylic acid methyl ester

  • MDL number MFCD01109140
  • Cas Number : 90087-77-7
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1411
Availability details for
Quantity
  • Availability
  • Add to Cart