COM448679383 Combi-Blocks, Inc.

4-Quinazolone-2-carboxylic Acid

  • MDL number MFCD02059339
  • Cas Number : 29113-34-6
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1417

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1417
Availability details for
Quantity
  • Availability
  • Add to Cart