COM448679390 Combi-Blocks, Inc.

3-Iodo-2,5,6-trimethoxypyridin-4-ol

  • MDL number MFCD22887386
  • Cas Number : 1414864-12-2
  • Molecular Weight : 311.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1427
Availability details for
Quantity
  • Availability
  • Add to Cart