COM448679392 Combi-Blocks, Inc.

Ethyl 4-benzyl-2-morpholinecarboxylate, HCl

  • MDL number MFCD18911733
  • Cas Number : 135782-26-2
  • Molecular Weight : 285.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1437

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1437
Availability details for
Quantity
  • Availability
  • Add to Cart