COM448679401 Combi-Blocks, Inc.

4-Morpholino-3-nitrobenzoic acid

  • MDL number MFCD00438745
  • Cas Number : 26577-59-3
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1449
Availability details for
Quantity
  • Availability
  • Add to Cart